Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminooctane, 98%

Catalog No. AAB2388514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2388514 25 g
AAB2388522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2388514 Supplier Thermo Scientific Chemicals Supplier No. B2388514
Only null left
Add to Cart

CAS: 373-44-4 | C8H22Cl2N2 | 217.18 g/mol

1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water 575g/L.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, carbon dioxide, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
IUPAC Name octane-1,8-diamine
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]

Specifications

Melting Point 50°C to 54°C
pH 11.7
Boiling Point 225°C to 226°C
Flash Point 106°C (223°F)
Odor Amine-like
Quantity 25 g
UN Number UN3259
Beilstein 1735426
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water 575g/L.
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 1,8-Diaminooctane
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 206-764-3
RTECSNumber RG8841500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.