Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminooctane, MP Biomedicals

Catalog No. ICN201951 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201951 10 g
ICN15759101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN201951 Supplier MP Biomedicals, Inc Supplier No. 0520195110

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
IUPAC Name octane-1,8-bis(aminium) dichloride
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]

Specifications

Quantity 10 g
Formula Weight 144.26
Chemical Name or Material 1, 8-Diaminooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.