Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,8-Diazabicyclo[5.4.0]Undec-7-Ene

Catalog No. 33482250MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-482-250MLF 250 mL
33-482-1LF 1 L
33-482-50MLF 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 33-482-250MLF Supplier Sigma Aldrich Supplier No. 33482250MLF
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,8-Diazabicyclo[5.4.0]undec-7-ene can be used:To mediate the aza-Michael addition of nitrogen nucleophiles to +a,+a-unsaturated carbonyl compounds.As a nucleophilic catalyst for the esterification of carboxylic acid with dimethyl carbonate.As a bidentate ligand in copper (I) bromide mediated atom transfer radical polymerization of methyl methacrylate.As an efficient catalyst for the transesterification of dimethyl carbonate with glycerol.Amidine base used for dehydrohalogenation reactions to olefins; Further important applications are e.g.: esterification of carboxylic acids with alkyl halides; Alkylation and acylation of active methylene compounds

Specifications

Density 1.019 g/mL at 20°C
Boiling Point 115°C (11 mmHg, lit.)
Molecular Formula C9H16N2
Linear Formula C9H16N2
Refractive Index n20/D 1.523
MDL Number MFCD00006930
Quantity 250 mL
Synonym 2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine; DBU
Molecular Weight (g/mol) 152.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.