Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,8-Diazabicyclo[5.4.0]Undec-7-Ene

Catalog No. 139009500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 x 2 g
25 g
100 g
500 g
2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-900-9500G 500 g
13-900-925KG 2.5 kg
13-900-925G 25 g
13-900-9100G 100 g
13-900-92X2G 2 x 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 13-900-9500G Supplier Sigma Aldrich Supplier No. 139009500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.Used in a new synthesis of the ABCD ring system of Camptothecin.

Specifications

Density 1.018 g/mL at 25°C
Boiling Point 80°C to 83°C (0.6 mmHg, lit.)
Molecular Formula C9H16N2
Refractive Index n20/D 1.522-1.524
Linear Formula C9H16N2
MDL Number MFCD00006930
Quantity 500 g
Synonym 2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine; DBU
Molecular Weight (g/mol) 152.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.