Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Dichlorooctane, 98%, Thermo Scientific™

Catalog No. AC255940250
Change view
Click to view available options
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC255940250 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255940250 Supplier Thermo Scientific Chemicals Supplier No. 255940250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2162-99-4
CAS Min % 97.5
CAS Max % 100.0
Density 1.02g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H16Cl2
Refractive Index 1.4580 to 1.4600 (20°C, 589nm)
Linear Formula Cl(CH2)8Cl
Quantity 25mL
MDL Number MFCD00039393
Synonym octane, 1,8-dichloro, 1,8-dichloroethane, octamethylene chloride, octamethylene dichloride, acmc-209fmk, 1,8-dichlorooctane
InChI Key WXYMNDFVLNUAIA-UHFFFAOYSA-N
SMILES ClCCCCCCCCCl
IUPAC Name 1,8-dichlorooctane
Molecular Weight (g/mol) 183.12
PubChem CID 75102
Formula Weight 183.12
Percent Purity ≥97.5%
Chemical Name or Material 1, 8-Dichlorooctane
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.