Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,8-dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione | 481-72-1 | MFCD00017373 | 270.24 | C15H10O5 | 10 MG
SDP

Catalog No. 5000295829
Encompass_Preferred

Aloe emodin (standard) is an analytical reference material intended for research and analytical applications, supplied with documentation for purity and used as a reference in chromatographic and mass-spectrometry methods for qualitative and quantitative analyses.

  • Molecular formula: C15H10O5.
  • Molecular weight: 270.24.
  • CAS number: 481-72-1.
  • Appearance: light brown to brown solid.
  • Intended use: analytical standard for HPLC, GC, and MS method development and validation.
  • Packaging options: available in 5 MG, 10 MG, 25 MG, 50 MG, 100 MG, 200 MG (quote), 500 MG (quote).

Catalog No. 50-002-95829 Supplier Medchemexpress LLC Supplier No. HYN0189R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aloe emodin (standard) is an analytical reference material intended for research and analytical applications, supplied with documentation for purity and used as a reference in chromatographic and mass-spectrometry methods for qualitative and quantitative analyses.

  • Molecular formula: C15H10O5.
  • Molecular weight: 270.24.
  • CAS number: 481-72-1.
  • Appearance: light brown to brown solid.
  • Intended use: analytical standard for HPLC, GC, and MS method development and validation.
  • Packaging options: available in 5 MG, 10 MG, 25 MG, 50 MG, 100 MG, 200 MG (quote), 500 MG (quote).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.