Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diphenyl-1,3,5,7-octatetraene, 98%, Thermo Scientific™

Catalog No. AC117360100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117360100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC117360100 Supplier Thermo Scientific Chemicals Supplier No. 117360100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3029-40-1
Molecular Formula C20H18
Molecular Weight (g/mol) 258.364
InChI Key ZENGMMQJMCPHTK-FPPPDJHPSA-N
Synonym 1,8-diphenyl-1,3,5,7-octatetraene, 1,8-diphenylocta-1,3,5,7-tetraene, benzene, 1,1'-1,3,5,7-octatetraene-1,8-diyl bis, benzene, 1,1'-1,3,5,7-octatetraene-1,8-diyl bis-, all-e, 1,8-diphenyloctatetraene, 1e,3e,5e,7e-8-phenylocta-1,3,5,7-tetraen-1-yl benzene, .alpha.,.omega.-diphenyloctatetraene, 1,1'-octa-1,3,5,7-tetraene-1,8-diyldibenzene, 8-phenylocta-1,3,5,7-tetraen-1-yl benzene, 1,3,5,7-octatetraene, 1,8-diphenyl
PubChem CID 5377501
IUPAC Name [(1E,3E,5E,7E)-8-phenylocta-1,3,5,7-tetraenyl]benzene
SMILES C1=CC=C(C=C1)C=CC=CC=CC=CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH=CHCH=CHCH=CHCH=CHC6H5
Quantity 10 g
Beilstein 05, 709
Formula Weight 258.35
Chemical Name or Material 1,8-Diphenyl-1,3,5,7-octatetraene, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.