Learn More
MSC CPC Colder Products MC2202 1/8" Inside Tube Diam, Brass, Quick Disconnect, Hose Barb Inline Coupling Insert 250 Max psi, -40 to 180°F, 1.12" Overall Length x 0.63" Overall Height, Chrome Plated
Catalog No. NC1706258
Encompass
CPC Colder Products MC2202 1/8" Inside Tube Diam, Brass, Quick Disconnect, Hose Barb Inline Coupling Insert 250 Max psi, -40 to 180°F, 1.12" Overall Length x 0.63" Overall Height, Chrome Plated
Catalog No. NC1706258 Supplier MSC Supplier No. 96295316
CPC Colder Products MC2202 1/8" Inside Tube Diam, Brass, Quick Disconnect, Hose Barb Inline Coupling Insert 250 Max psi, -40 to 180°F, 1.12" Overall Length x 0.63" Overall Height, Chrome Plated