Learn More
Chemglass Life Sciences 1/8" Hose Barb Non-Valved In-Line Coupling Body, Polysulfone, CPC MPC17002T39
SDP
Catalog No. 5000980608
Encompass
MPC Series couplings add ease of use and security to your most critical fluid handling applications. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, in sizes to fit 1/8", 1/4" and 3/8" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.Features: Ergonomic thumb latch. USP Class VI materials. Sterilizable by autoclave, Et0, e-beam or gamma. Parting line-free hose barb. BSE/TSE free materials   Benefits: Easy to operate – even with gloved hands. Meet biocompatibility requirements. Reusable, yet economical enough to allow disposability. Eliminates potential leak path. Meet ADCF requirements.
Catalog No. 50-009-80608 Supplier Chemglass Life Sciences Supplier No. CLS91402039
MPC Series couplings add ease of use and security to your most critical fluid handling applications. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, in sizes to fit 1/8", 1/4" and 3/8" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.Features: Ergonomic thumb latch. USP Class VI materials. Sterilizable by autoclave, Et0, e-beam or gamma. Parting line-free hose barb. BSE/TSE free materials   Benefits: Easy to operate – even with gloved hands. Meet biocompatibility requirements. Reusable, yet economical enough to allow disposability. Eliminates potential leak path. Meet ADCF requirements.