Learn More
Electron Microscopy Sciences Pro-Craft 1/8 Hp Flex Shaft Machine, 110 V
SDP
Catalog No. 50996784
Encompass_Preferred
A versatile hang-up flexible shaft machine. Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 - 18 RPM. Complete kit comes with a hand piece, 39" (1 m) Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Catalog No. 50-996-784 Supplier Electron Microscopy Sciences Supplier No. 6213010
A versatile hang-up flexible shaft machine. Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 - 18 RPM. Complete kit comes with a hand piece, 39" (1 m) Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.