Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, Food-Grade, 1/8 in ID, 50 ft
Catalog No. NC3660103
Encompass_Preferred
Saint-Gobain Tygon S3 B-44-4X tubing is a clear, flexible, food- and beverage-grade transfer tubing designed for sanitary fluid handling in processing and dispensing systems. It features a smooth, non-porous bore that minimizes particle entrapment and resists common sanitizers and alkaline cleaners, making it suitable for dairy, condiment, and beverage applications.
  • Food-grade, phthalate-free formulation compliant with FDA and NSF food-contact standards.
  • 1/8 in (3.18 mm) inner diameter, 3/16 in (4.76 mm) outer diameter, 1/32 in (0.79 mm) wall thickness.
  • Sold in 50 ft (15 m) roll for continuous runs and easy installation.
  • Clear, flexible construction with a smooth bore for visual flow monitoring and sanitary transfer.
  • Maximum working pressure 34 psi (2.3 bar) at 73°F (23°C); vacuum rating 25.0 inHg (635 mmHg).
  • Operating temperature range -49°F (-45°C) to 165°F (74°C).
Catalog No. NC3660103 Supplier Accuflow Systems Supplier No. AAA00006
Saint-Gobain Tygon S3 B-44-4X tubing is a clear, flexible, food- and beverage-grade transfer tubing designed for sanitary fluid handling in processing and dispensing systems. It features a smooth, non-porous bore that minimizes particle entrapment and resists common sanitizers and alkaline cleaners, making it suitable for dairy, condiment, and beverage applications.
  • Food-grade, phthalate-free formulation compliant with FDA and NSF food-contact standards.
  • 1/8 in (3.18 mm) inner diameter, 3/16 in (4.76 mm) outer diameter, 1/32 in (0.79 mm) wall thickness.
  • Sold in 50 ft (15 m) roll for continuous runs and easy installation.
  • Clear, flexible construction with a smooth bore for visual flow monitoring and sanitary transfer.
  • Maximum working pressure 34 psi (2.3 bar) at 73°F (23°C); vacuum rating 25.0 inHg (635 mmHg).
  • Operating temperature range -49°F (-45°C) to 165°F (74°C).