Learn More
Waters Corp Tubing, 0.125 in OD, 0.062 in ID, 120.00 in Length, Stainless Steel
Catalog No. 50785257 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Tubing and fittings are designed to provide leak-free connections with low dead volume and minimal band spreading. For optimal performance, these components must be clean and designed to work together. The geometry of the ferrule and the conical port must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths, and diameters are available to provide flexibility when setting up and maintaining a system.
  • Designed to provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Components must be clean and designed to work together for best performance
  • Geometry of the ferrule and conical port must match
  • Diameter of the tube and fittings must match
  • Available in a variety of materials, lengths, and diameters
  • Made of stainless steel
Catalog No. 50-785-257 Supplier Waters Corp Supplier No. WAT026806
Tubing and fittings are designed to provide leak-free connections with low dead volume and minimal band spreading. For optimal performance, these components must be clean and designed to work together. The geometry of the ferrule and the conical port must match, as well as the diameter of the tube and the fittings. A variety of materials, lengths, and diameters are available to provide flexibility when setting up and maintaining a system.
  • Designed to provide leak-free connections
  • Low dead volume
  • Minimal band spreading
  • Components must be clean and designed to work together for best performance
  • Geometry of the ferrule and conical port must match
  • Diameter of the tube and fittings must match
  • Available in a variety of materials, lengths, and diameters
  • Made of stainless steel