Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Naphthalimide 98.0+%, TCI America™
SDP

Catalog No. N045625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N045625G 25 g
N0456250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N045625G Supplier TCI America Supplier No. N045625G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 81-83-4
Molecular Formula C12H7NO2
Molecular Weight (g/mol) 197.193
MDL Number MFCD00006920
InChI Key XJHABGPPCLHLLV-UHFFFAOYSA-N
Synonym 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn
PubChem CID 66491
IUPAC Name benzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2

Specifications

Melting Point 301°C
Quantity 25 g
Formula Weight 197.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Naphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.