Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Naphthosultam 98%, Thermo Scientific™

Catalog No. AC211351000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC211351000 100 g
AC211350250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211351000 Supplier Thermo Scientific Chemicals Supplier No. 211351000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-72-5
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
InChI Key HOBYPSNNFBRLIX-UHFFFAOYSA-N
Synonym 1,8-naphthosultam, 1,8-naphthalenesultam, 2h-naphtho 1,8-cd isothiazole 1,1-dioxide, 2h-naphth 1,8-cd isothiazole 1,1-dioxide, 2h-naphth 1,8-cd isothiazole,1,1-dioxide, 2??-thia-3-azatricyclo 6.3.1.0?, 1 2 dodeca-1 11 ,4 12 ,5,7,9-pentaene-2,2-dione, 1,8-naphthasultam, naphtho 1,8-cd isothiazole 1,1-dioxide
PubChem CID 69052
SMILES C1=CC2=C3C(=C1)NS(=O)(=O)C3=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 175°C to 179°C
Color Brown-Gray to Gray-Green or Gray
Infrared Spectrum Authentic
Quantity 100 g
Beilstein 27, 59
Formula Weight 205.23
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 1, 8-Naphthosultam, 98%
Show More Show Less
EINECSNumber 210-055-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.