Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,8-naphthyridine-1(2H)-carboxamide, 3,4-dihydro-6-[5-(methoxymethyl)-3-pyridinyl]- | 1633009-87-6 | 99.2% | 298.34 g·mol-1 | C16H18N4O2 | 1 ML
SDP

Catalog No. 5000212426
Encompass_Preferred

BI 689648 is a small-molecule, highly selective aldosterone synthase (CYP11B2) inhibitor used in preclinical research. It potently inhibits CYP11B2 while showing substantially less activity against CYP11B1, and is supplied with molecular characterization, purity data, and solubility/formulation guidance for in vitro and in vivo studies.

  • Potent aldosterone synthase inhibition (IC50 2.1 nM).
  • Reduced activity against CYP11B1 (IC50 310 nM) for improved selectivity.
  • Characterized molecular data (C16H18N4O2; MW 298.34 g·mol-1).
  • High purity suitable for research use (99.2% by LCMS).
  • Documented solubility and formulation guidance for DMSO and in vivo vehicles.

Catalog No. 50-002-12426 Supplier Medchemexpress LLC Supplier No. HY10121710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI 689648 is a small-molecule, highly selective aldosterone synthase (CYP11B2) inhibitor used in preclinical research. It potently inhibits CYP11B2 while showing substantially less activity against CYP11B1, and is supplied with molecular characterization, purity data, and solubility/formulation guidance for in vitro and in vivo studies.

  • Potent aldosterone synthase inhibition (IC50 2.1 nM).
  • Reduced activity against CYP11B1 (IC50 310 nM) for improved selectivity.
  • Characterized molecular data (C16H18N4O2; MW 298.34 g·mol-1).
  • High purity suitable for research use (99.2% by LCMS).
  • Documented solubility and formulation guidance for DMSO and in vivo vehicles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.