Learn More
Medchemexpress LLC 1,8-Octanediol | 629-41-4 | MFCD00002989 | 100.0% | 146.23 g/mol | C8H18O2 | 100 G
SDP
Catalog No. 5000300189
Encompass_Preferred
1,8-Octanediol is a straight-chain aliphatic diol (C8) used as a building block in polymer synthesis, elastomer formation, and surface modification. It is supplied as a white to off-white solid for research applications and offers high chemical purity suitable for laboratory-scale synthesis and formulation work.
  • High purity for research and synthesis applications.
  • Serves as a diol monomer for polyesters and polyurethanes.
  • Useful as a spacer in elastomer and hybrid material synthesis.
  • Solid form simplifies handling and storage.
  • Well-characterized: chemical formula C8H18O2, molecular weight 146.23 g/mol.
Catalog No. 50-003-00189 Supplier Medchemexpress LLC Supplier No. HYW017508100G
1,8-Octanediol is a straight-chain aliphatic diol (C8) used as a building block in polymer synthesis, elastomer formation, and surface modification. It is supplied as a white to off-white solid for research applications and offers high chemical purity suitable for laboratory-scale synthesis and formulation work.
  • High purity for research and synthesis applications.
  • Serves as a diol monomer for polyesters and polyurethanes.
  • Useful as a spacer in elastomer and hybrid material synthesis.
  • Solid form simplifies handling and storage.
  • Well-characterized: chemical formula C8H18O2, molecular weight 146.23 g/mol.