Learn More
Medchemexpress LLC 1,8-octanediol | 629-41-4 | MFCD00002989 | 99.8% | 146.23 g/mol | C8H18O2 | 50 G
SDP
Catalog No. 5000300413
Encompass_Preferred
1,8-Octanediol is an aliphatic diol (C8H18O2) used as a biochemical reagent and materials building block. It is applied in membrane modification to improve hemocompatibility, in synthesis of hybrid elastomers for soft and hard tissue regeneration, and as a reagent in biochemical assays. Available as a high-purity powder suitable for research and materials synthesis.
  • High purity (99.8% GC) for analytical and synthetic applications.
  • Low water content (0.01% KF) for moisture-sensitive reactions.
  • Powder form, facilitating handling and formulation.
  • Useful for membrane modification to enhance hemocompatibility.
  • Serves as a building block for hybrid elastomers and polymer synthesis.
  • Molecular weight 146.23 g/mol; formula C8H18O2.
Catalog No. 50-003-00413 Supplier Medchemexpress LLC Supplier No. HYW01750850G
1,8-Octanediol is an aliphatic diol (C8H18O2) used as a biochemical reagent and materials building block. It is applied in membrane modification to improve hemocompatibility, in synthesis of hybrid elastomers for soft and hard tissue regeneration, and as a reagent in biochemical assays. Available as a high-purity powder suitable for research and materials synthesis.
  • High purity (99.8% GC) for analytical and synthetic applications.
  • Low water content (0.01% KF) for moisture-sensitive reactions.
  • Powder form, facilitating handling and formulation.
  • Useful for membrane modification to enhance hemocompatibility.
  • Serves as a building block for hybrid elastomers and polymer synthesis.
  • Molecular weight 146.23 g/mol; formula C8H18O2.