Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Octanediol, 98%

Catalog No. AC129381000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC129381000 100 g
AC129380250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129381000 Supplier Thermo Scientific Chemicals Supplier No. 129381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 629-41-4 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-41-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
InChI Key OEIJHBUUFURJLI-UHFFFAOYSA-N
PubChem CID 69420
ChEBI CHEBI:44630
IUPAC Name octane-1,8-diol
SMILES C(CCCCO)CCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 60.0°C to 62.0°C
Color White to Yellow
Boiling Point 278.8°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HO(CH2)8OH
Quantity 100 g
Beilstein 01, 490
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Solubility Information Solubility in water: slightly soluble in water.
Viscosity 20.6 mPa.s (70°C)
Formula Weight 146.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1, 8-Octanediol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 211-090-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.