Learn More
Glass Expansion 1/8" Tefzel Ferrules (PKT 10) for FT-16-8-X
SDP
Catalog No. 502703049
Encompass_Preferred
1/8” Tefzel Ferrules (PKT 10). Package of ten replacement flangeless Tefzel ferrules 1/8”. Compatible with Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8-X. For 1.6mm (1/16”) Tefzel Ferrules see 70-803-1315. Note: For Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8 use 1/8” Tefzel Flangeless Ferrule 70-803-1316.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3049 Supplier Glass Expansion Supplier No. 708031513
1/8” Tefzel Ferrules (PKT 10). Package of ten replacement flangeless Tefzel ferrules 1/8”. Compatible with Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8-X. For 1.6mm (1/16”) Tefzel Ferrules see 70-803-1315. Note: For Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8 use 1/8” Tefzel Flangeless Ferrule 70-803-1316.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001