Learn More
Glass Expansion 1/8" Tefzel Ferrules (PKT 10) for FT-16-8
SDP
Catalog No. 502703062
Encompass_Preferred
1/8” Tefzel Ferrules (PKT 10). Package of ten replacement flangeless 1/8” Tefzel ferrules. Compatible with Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8. For 1.6mm (1/16”) Tefzel Ferrules see 70-803-1315. Note: For Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8-X use 1/8” Tefzel Flangeless Ferrule 70-803-1513.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3062 Supplier Glass Expansion Supplier No. 708031316
1/8” Tefzel Ferrules (PKT 10). Package of ten replacement flangeless 1/8” Tefzel ferrules. Compatible with Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8. For 1.6mm (1/16”) Tefzel Ferrules see 70-803-1315. Note: For Glass Expansion's High Pressure Nebulizer Interface Kit FT-16-8-X use 1/8” Tefzel Flangeless Ferrule 70-803-1513.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001