Learn More
Waters Corp Waters Corporation Tubing, Stainless Steel, 1/8 in. OD, 0.085 in. ID
Catalog No. 50466717 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This stainless steel tubing serves as a quality spare part for various laboratory applications. It features an outer diameter of 1/8 in. and an inner diameter of 0.085 in., with a length of 5 m. This tubing is designed to ensure reliable performance and easy replacement.
  • Quality spare part.
  • Designed as a spare part kit.
  • One unit per package.
  • Replacement as required.
Catalog No. 50-466-717 Supplier Waters Corp Supplier No. 6395995S5
This stainless steel tubing serves as a quality spare part for various laboratory applications. It features an outer diameter of 1/8 in. and an inner diameter of 0.085 in., with a length of 5 m. This tubing is designed to ensure reliable performance and easy replacement.
  • Quality spare part.
  • Designed as a spare part kit.
  • One unit per package.
  • Replacement as required.