Learn More
MSC CPC Colder Products PMC220112 1/8" Nominal Flow, 1/16" ID, Male, Inline Hose Barb-Male Plug 120 Max psi, 32 to 160°F, Polypropylene Coupling, EPDM O-Ring
Catalog No. NC1706260
Encompass
CPC Colder Products PMC220112 1/8" Nominal Flow, 1/16" ID, Male, Inline Hose Barb-Male Plug 120 Max psi, 32 to 160°F, Polypropylene Coupling, EPDM O-Ring
Catalog No. NC1706260 Supplier MSC Supplier No. 48731301
CPC Colder Products PMC220112 1/8" Nominal Flow, 1/16" ID, Male, Inline Hose Barb-Male Plug 120 Max psi, 32 to 160°F, Polypropylene Coupling, EPDM O-Ring