Learn More
Ace Glass, Inc. PTFE tubing, 1/8in OD x 312mm, connect to solvent reservoir and feed up through the sample tube to be cleaned
SDP
Catalog No. 50971333
Encompass_Preferred
1/8 O.D. x 312mm PTFE tubing, for use with NMR tube washer. Tubing connects to the solvent reservoir and feeds up through the sample tube to be cleaned.
Catalog No. 50-971-333 Supplier Ace Glass, Inc. Supplier No. 254031
1/8 O.D. x 312mm PTFE tubing, for use with NMR tube washer. Tubing connects to the solvent reservoir and feeds up through the sample tube to be cleaned.