Learn More
BRANSON ULTRASONICS CORPORATION (3P) 1/8IN DOUBLESTP MT 500/250/450
Catalog No. NC3721569
Encompass_Preferred
Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers, 1/8 in. double-stepped microtip, Amplitude Range: 64 to 247um, Diameter Tip: 0.12 in., 0.3cm, Intensity: Very high, Tip Style: Double stepped, Capacity: 0.1 to 10mL
Catalog No. NC3721569 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101063212
Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers, 1/8 in. double-stepped microtip, Amplitude Range: 64 to 247um, Diameter Tip: 0.12 in., 0.3cm, Intensity: Very high, Tip Style: Double stepped, Capacity: 0.1 to 10mL