Learn More
BRANSON ULTRASONICS CORPORATION (3P) 1/8IN MICROTIP 500/550/250/450
Catalog No. NC3721566
Encompass_Preferred
Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers, 1/8 in. tapered tip, Amplitude Range: 116 to 494um, Diameter Tip: 0.12 in., 0.3cm, Intensity: Ultra high, Tip Style: Tapered, Capacity: 0.25 to 10mL
Catalog No. NC3721566 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101148062
Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers, 1/8 in. tapered tip, Amplitude Range: 116 to 494um, Diameter Tip: 0.12 in., 0.3cm, Intensity: Ultra high, Tip Style: Tapered, Capacity: 0.25 to 10mL