Learn More
Sonics & Materials Inc 1/8IN TAPERED MICROTIP
SDP
Catalog No. NC9841812
Encompass_Preferred
Tapered microtip 0.125 in. (3mm),compatible with most 20 kHz Ultrasonic Processors,Do not use tapered microtip with coupler,TIP DIAMETER: 0.125 in. (3mm),INTENSITY: Ultra High ,VOLUME (batch): 1-10mL,AMPLITUDE:228mm / .0090 in. ,Fits 5mL te
Catalog No. NC9841812 Supplier Sonics & Materials Inc Supplier No. 6300418
Tapered microtip 0.125 in. (3mm),compatible with most 20 kHz Ultrasonic Processors,Do not use tapered microtip with coupler,TIP DIAMETER: 0.125 in. (3mm),INTENSITY: Ultra High ,VOLUME (batch): 1-10mL,AMPLITUDE:228mm / .0090 in. ,Fits 5mL te