Learn More
Cole-Parmer Threaded Ultrasonic Probe Tip, Stepped; 1/8" (3mm) Dia
Supplier:  Cole-Parmer 0471212
Encompass_Preferred
Probes are made of titanium and are autoclavable. Optional 2-mm probe fits into microcentrifuge tubes; use to process small-volume samples.
Catalog No. NC2128590