Learn More
Ace Glass, Inc. Heating tape, 1in, 6ft length, 300w at 230v max, Glas-Col; 103A SET 16
SDP
Catalog No. 50983299
Encompass
1 Glas-Col heating tape eliminates the need for a reducing voltage transformer. 6 foot length. Can be operated directly through a temperature control at normal outlet voltage. 300w at 230v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-983-299 Supplier Ace Glass, Inc. Supplier No. 1206348
1 Glas-Col heating tape eliminates the need for a reducing voltage transformer. 6 foot length. Can be operated directly through a temperature control at normal outlet voltage. 300w at 230v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.