Learn More
CELLTREAT Scientific Products 1.8mL CF Cryogenic Vial, Internal Thread, Self-Standing, Sterile, 500/cs
SDP
Catalog No. NC3433482
Encompass_Preferred
"CELLTREAT Freeze Cryogenic Vials are designed for storing biological material at temperatures as low as -196°C, in the gas phase of liquid nitrogenAvailable in round bottom or self-standing optionsCELLTREAT product descriptions are based on the highest graduation mark on the vial. Other brands may name vials based on actual liquid capacityRe-sealable bags offer a convenient storage solutionInternal thread cap prevents contamination and uses O-ring silicone seal to ensure positive seal at any temperatureCaps have textured ridges to provide gripping when wearing glovesContrasting black graduations are large and easy to viewLarge white marking area for writing also provides backdrop to enhance black graduationsSelf-standing base interlocks with Easy Grip Workstation Rack for easy one-handed opening"
Catalog No. NC3433482 Supplier CELLTREAT Scientific Products Supplier No. 229915B
"CELLTREAT Freeze Cryogenic Vials are designed for storing biological material at temperatures as low as -196°C, in the gas phase of liquid nitrogenAvailable in round bottom or self-standing optionsCELLTREAT product descriptions are based on the highest graduation mark on the vial. Other brands may name vials based on actual liquid capacityRe-sealable bags offer a convenient storage solutionInternal thread cap prevents contamination and uses O-ring silicone seal to ensure positive seal at any temperatureCaps have textured ridges to provide gripping when wearing glovesContrasting black graduations are large and easy to viewLarge white marking area for writing also provides backdrop to enhance black graduationsSelf-standing base interlocks with Easy Grip Workstation Rack for easy one-handed opening"