Learn More
Ace Glass, Inc. Heating tape, 5/8in, 6ft length, 120w at 115v max, Glas-Col;103A FET0.66
SDP
Catalog No. 50983297
Encompass
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 6 foot length. Can be operated directly through a temperature control at normal outlet voltage. 120w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-983-297 Supplier Ace Glass, Inc. Supplier No. 1206304
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 6 foot length. Can be operated directly through a temperature control at normal outlet voltage. 120w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.