Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Diacetoxynonane 98.0+%, TCI America™
SDP

Catalog No. N062325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N062325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N062325G Supplier TCI America Supplier No. N062325G

Chemical Identifiers

CAS 4944-60-9
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
MDL Number MFCD00450895
InChI Key JAEVCKMMGIFOCL-UHFFFAOYSA-N
Synonym Nonamethylene Diacetate, 1,9-Nonanediol Diacetate
PubChem CID 243677
IUPAC Name 9-(acetyloxy)nonyl acetate
SMILES CC(=O)OCCCCCCCCCOC(C)=O

Specifications

Color Colorless
Boiling Point 161°C
Quantity 25 g
Formula Weight 244.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,9-Diacetoxynonane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.