Learn More
Medchemexpress LLC 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azatridecan-13-oic acid | 872679-70-4 | MFCD08064306 | 99.8% | 399.44 g/mol | C22H25NO6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0402381G
Encompass_Preferred
Fmoc-NH-PEG2-CH2CH2COOH is an Fmoc-protected PEG2 linker with a terminal carboxylic acid, used as a building block in bioconjugation chemistry including ADC and PROTAC synthesis. The Fmoc group protects the amine during multistep synthesis and can be removed under standard deprotection conditions.
  • Fmoc-protected amine for selective protection during synthesis
  • Terminal carboxylic acid functionality for coupling reactions
  • PEG2 spacer improves solubility and provides flexibility
  • High purity (99.8%) suitable for research synthesis
  • Molecular weight 399.44 g/mol; formula C22H25NO6
  • Available as a 1 g pack for small-scale synthesis
Catalog No. 50-002-03212