Learn More
Medchemexpress LLC 1-(9H-fluoren-9-ylmethoxycarbonyl)-4-[(2-methylpropan-2-yl)oxycarbonylamino]piperidine-4-carboxylic acid | 368866-07-3 | MFCD02683117 | 99.4% | 466.53 | C26H30N2O6 | 1 ML
SDP
Catalog No. 5000301454
Encompass_Preferred
SARS-CoV-2-IN-86 is a research-grade small molecule inhibitor of SARS-CoV-2 methyltransferases (nsp14 and nsp16), provided for in vitro biochemical and antiviral studies. It is available as solid material and as a 10 mM solution in DMSO, with documented purity and storage recommendations to support reproducible experimental use.
  • High purity (99.35%) for reproducible results.
  • Available as 10 mM solution in DMSO or as solid for flexible assay workflows.
  • Well-characterized chemical identity (CAS 368866-07-3) and formula for traceability.
  • Suitable for biochemical methyltransferase and antiviral research applications.
  • Defined storage conditions to preserve stability in powder and solution formats.
  • Datasheet available with specification and handling information.
Catalog No. 50-003-01454 Supplier Medchemexpress LLC Supplier No. HYW02448510MM/1ML
SARS-CoV-2-IN-86 is a research-grade small molecule inhibitor of SARS-CoV-2 methyltransferases (nsp14 and nsp16), provided for in vitro biochemical and antiviral studies. It is available as solid material and as a 10 mM solution in DMSO, with documented purity and storage recommendations to support reproducible experimental use.
  • High purity (99.35%) for reproducible results.
  • Available as 10 mM solution in DMSO or as solid for flexible assay workflows.
  • Well-characterized chemical identity (CAS 368866-07-3) and formula for traceability.
  • Suitable for biochemical methyltransferase and antiviral research applications.
  • Defined storage conditions to preserve stability in powder and solution formats.
  • Datasheet available with specification and handling information.