Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(9H-fluoren-9-ylmethoxycarbonyl)azetidine-3-carboxylic acid | 193693-64-0 | MFCD01321018 | 99.9% | 323.35 g/mol | C19H17NO4 | 5 G
SDP

Catalog No. 5000203073
Encompass_Preferred

Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected azetidine-3-carboxylic acid used as a building block in peptide synthesis and as a cleavable linker in antibody-drug conjugate construction. It is supplied at high reported purity for research use.

  • CAS number: 193693-64-0.
  • Purity: 99.9% (reported).
  • Molecular formula: C19H17NO4.
  • Molecular weight: 323.35 g/mol.
  • Common applications: peptide building block and ADC linker.
  • Available in laboratory-scale quantities, including 5 G packages.

Catalog No. 50-002-03073 Supplier Medchemexpress LLC Supplier No. HYW0112775G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-azetidine-3-carboxylic acid is an Fmoc-protected azetidine-3-carboxylic acid used as a building block in peptide synthesis and as a cleavable linker in antibody-drug conjugate construction. It is supplied at high reported purity for research use.

  • CAS number: 193693-64-0.
  • Purity: 99.9% (reported).
  • Molecular formula: C19H17NO4.
  • Molecular weight: 323.35 g/mol.
  • Common applications: peptide building block and ADC linker.
  • Available in laboratory-scale quantities, including 5 G packages.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.