Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(a-D-ribofuranosyl)uracil | 3258-07-9 | MFCD17430297 | 97.0% | 244.20 g/mol | C9H12N2O6 | 50 MG
SDP

Catalog No. 5000282623
Encompass_Preferred

1-(a-D-ribofuranosyl)uracil is a purine nucleoside analog for research use. It has reported antitumor activity, particularly against indolent lymphoid malignancies, and its mechanisms include inhibition of DNA synthesis and induction of apoptosis. The compound is supplied as a solid with defined purity and solubility profiles suitable for in vitro and in vivo studies.

  • Purity 97.0%.
  • Molecular weight 244.20 g/mol.
  • Chemical formula C9H12N2O6.
  • CAS number 3258-07-9.
  • Appearance white to off-white solid.
  • Package sizes include 50 mg, 100 mg, 250 mg.
  • Storage (powder) -20°C for 3 years or 4°C for 2 years.
  • Solubility in vitro: DMSO 100 mg/mL; requires ultrasonic.
  • In vivo formulations achieve ≥ 2.5 mg/mL using recommended protocols.

Catalog No. 50-002-82623 Supplier Medchemexpress LLC Supplier No. HYW12811250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-(a-D-ribofuranosyl)uracil is a purine nucleoside analog for research use. It has reported antitumor activity, particularly against indolent lymphoid malignancies, and its mechanisms include inhibition of DNA synthesis and induction of apoptosis. The compound is supplied as a solid with defined purity and solubility profiles suitable for in vitro and in vivo studies.

  • Purity 97.0%.
  • Molecular weight 244.20 g/mol.
  • Chemical formula C9H12N2O6.
  • CAS number 3258-07-9.
  • Appearance white to off-white solid.
  • Package sizes include 50 mg, 100 mg, 250 mg.
  • Storage (powder) -20°C for 3 years or 4°C for 2 years.
  • Solubility in vitro: DMSO 100 mg/mL; requires ultrasonic.
  • In vivo formulations achieve ≥ 2.5 mg/mL using recommended protocols.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.