Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1'-Acetonaphthone, 95%

Catalog No. AC400090010
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400090010 1 kg
AC400092500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400090010 Supplier Thermo Scientific Chemicals Supplier No. 400090010
Add to Cart

CAS: 941-98-0 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 12.0°C
Color Yellow
Density 1.1190g/mL
Boiling Point 302.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6270 to 1.6290
Linear Formula C10H7COCH3
Quantity 1 kg
Specific Gravity 1.119
Solubility Information Solubility in water: immiscible
Formula Weight 170.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1'-Acetonaphthone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-384-1
RTECSNumber AL2982800
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.