Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetoxy-2-methoxy-4-[(E)-1-propenyl]benzene 98.0+%, TCI America™
SDP

Catalog No. A1184500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1184500G 500 g
A118425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1184500G Supplier TCI America Supplier No. A1184500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93-29-8
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00026984
InChI Key IUSBVFZKQJGVEP-SNAWJCMRSA-N
Synonym Acetylisoeugenol
PubChem CID 876160
ChEBI CHEBI:86583
IUPAC Name 2-methoxy-4-[(1E)-prop-1-en-1-yl]phenyl acetate
SMILES COC1=CC(\C=C\C)=CC=C1OC(C)=O

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 283°C
Quantity 500 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetoxy-2-methoxy-4-[(E)-1-propenyl]benzene
RTECSNumber SL7940000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.