Learn More
Cayman Chemical 1β-Acetoxypolygodial, Purity: ≥95%, CAS: 1818425-44-3, 5 mg
SDP
Catalog No. 502992397
Encompass_Preferred
A sesquiterpenoid; active against T. cruzi amastigotes and trypomastigotes (GI50s = 57.8 and 72 µM, respectively) but not T. cruzi epimastigotes (GI50 = >100 µM)
Catalog No. 50-299-2397 Supplier Cayman Chemical Supplier No. 299015 MG
A sesquiterpenoid; active against T. cruzi amastigotes and trypomastigotes (GI50s = 57.8 and 72 µM, respectively) but not T. cruzi epimastigotes (GI50 = >100 µM)