Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-1,2,3,4-tetrahydroquinoline 98.0+%, TCI America™
SDP

Catalog No. A16415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16415G 5 g
A164125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16415G Supplier TCI America Supplier No. A16415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4169-19-1
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD01195305
InChI Key RRWLNRQGJSQRAF-UHFFFAOYSA-N
Synonym 1,2,3,4-Tetrahydro-1-acetylquinoline
PubChem CID 347850
IUPAC Name 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one
SMILES CC(=O)N1CCCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 295°C
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetyl-1,2,3,4-tetrahydroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.