Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Acetyl-5-bromo-1H-indole

Catalog No. VBP001931G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP001931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP001931G Supplier Sigma Aldrich Supplier No. VBP001931G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Electronic structure of the lowest excited triplet state of 6,13-pentacenequinone has been studied by continuous-wave time-resolved electron paramagnetic resonance and pulsed electron nuclear double resonance.6,13-pentacenequinone has been used in:
  • Deposition of 6,13-pentacenequinone thin films on n-si substrates by thermal evaporation
  • As precursor to diarylpentacenes, potential organic field-effect transitors

Specifications

CAS 61995-52-6
Molecular Formula C10H8BrNO
Linear Formula C10H8BrNO
MDL Number MFCD00238530
Quantity 1 g
Molecular Weight (g/mol) 238.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.