Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylferrocene, 99+%, Thermo Scientific™

Catalog No. AA2295406
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA2295406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA2295406 Supplier Thermo Scientific Chemicals Supplier No. 2295406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1271-55-2
Molecular Formula C12H12FeO
Quantity 5g
MDL Number MFCD00001432
UN Number UN3467
Synonym Acetylferrocene; [(Acetylcyclopentadienyl)cyclopentadienyliron]
InChI Key SPKJCVZOZISLEI-UHFFFAOYSA-N
Molecular Weight (g/mol) 228.07
Formula Weight 228.07
Percent Purity ≥99%
Chemical Name or Material 1-Acetylferrocene
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H310
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
P280-P301+P310-P361-P302+P350-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.