Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acryloyloxy-3-hydroxyadamantane 98.0+%, TCI America™
SDP

Catalog No. A28595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28595G 5 g
A285925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28595G Supplier TCI America Supplier No. A28595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 216581-76-9
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD13194929
InChI Key DKDKCSYKDZNMMA-UHFFFAOYSA-N
Synonym 3-Hydroxy-1-adamantyl Acrylate, Acrylic Acid 3-Hydroxy-1-adamantyl Ester
PubChem CID 22228790
IUPAC Name (3-hydroxy-1-adamantyl) prop-2-enoate
SMILES C=CC(=O)OC12CC3CC(C1)CC(C3)(C2)O

Specifications

Melting Point 72°C
Color White
Quantity 5 g
Formula Weight 222.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acryloyloxy-3-hydroxyadamantane
EINECSNumber (4)-1910
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.