Learn More
Medchemexpress LLC 1-Adamantan-d15-amine | 33830-10-3 | MFCD01317436 | 98.0% | 166.34 | C10H2D15N | 10 MG
SDP
Catalog No. 5000298096
Encompass_Preferred
Amantadine-d15 (1-Adamantanamine-d15) is the deuterium-labeled isotopologue of amantadine supplied as a solid analytical standard for use in mass spectrometry, isotope-dilution quantitation, metabolic, and pharmacokinetic studies. The product is characterized by its molecular formula and verified purity on the certificate of analysis, and is intended for use as an internal standard or tracer in quantitative workflows.
  • High isotopic labeling suitable for mass spectrometry applications.
  • Purity 98.0% as reported on the certificate of analysis.
  • White to off-white solid for convenient storage and handling.
  • Appropriate as an internal standard for LC-MS/MS and GC-MS quantitation.
  • Provided in a 10 mg quantity for analytical use.
Catalog No. 50-002-98096 Supplier Medchemexpress LLC Supplier No. HYB0402S10MG
Add to Cart
Add to Cart
Amantadine-d15 (1-Adamantanamine-d15) is the deuterium-labeled isotopologue of amantadine supplied as a solid analytical standard for use in mass spectrometry, isotope-dilution quantitation, metabolic, and pharmacokinetic studies. The product is characterized by its molecular formula and verified purity on the certificate of analysis, and is intended for use as an internal standard or tracer in quantitative workflows.
  • High isotopic labeling suitable for mass spectrometry applications.
  • Purity 98.0% as reported on the certificate of analysis.
  • White to off-white solid for convenient storage and handling.
  • Appropriate as an internal standard for LC-MS/MS and GC-MS quantitation.
  • Provided in a 10 mg quantity for analytical use.