Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine Hydrochloride 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
665-66-7
C10H17N·HCl
187.71
MFCD00074723
WOLHOYHSEKDWQH-UHFFFAOYSA-N
amantadine hydrochloride,1-adamantanamine hydrochloride,symmetrel,1-aminoadamantane hydrochloride,amantadine hcl,adamantanamine hydrochloride,amazolon,midantan,midantane,mydantane
64150
CHEBI:2619
adamantan-1-amine;hydrochloride
C1C2CC3CC1CC(C2)(C3)N.Cl
665-66-7
C10H17N·HCl
187.71
MFCD00074723
WOLHOYHSEKDWQH-UHFFFAOYSA-N
amantadine hydrochloride,1-adamantanamine hydrochloride,symmetrel,1-aminoadamantane hydrochloride,amantadine hcl,adamantanamine hydrochloride,amazolon,midantan,midantane,mydantane
64150
CHEBI:2619
adamantan-1-amine;hydrochloride
C1C2CC3CC1CC(C2)(C3)N.Cl
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Melting Point >300.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Merck Index 15, 368
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
Formula Weight 187.71
Physical Form Fine Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material 1-Adamantanamine hydrochloride, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-560-2

RTECSNumber : AU4375000

TSCA : TSCA