Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine hydrochloride, 99+%

Catalog No. AC151671000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC151671000 100 g
AC151670250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151671000 Supplier Thermo Scientific Chemicals Supplier No. 151671000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 665-66-7 | C10H18ClN | 187.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
Synonym amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
PubChem CID 64150
ChEBI CHEBI:2619
IUPAC Name adamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 99
CAS Max % 100
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 368
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
Formula Weight 187.71
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Adamantanamine hydrochloride, 99+%
Show More Show Less
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed
EINECSNumber 211-560-2
RTECSNumber AU4375000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.