Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. A131925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A131925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A131925G Supplier TCI America Supplier No. A131925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31377-23-8
Molecular Formula C10H17N
MDL Number MFCD00077251
Synonym 1-adamantanamine sulfate, adamantan-1-amine sulfate, 1-aminoadamantane sulfate, tricyclo 3.3.1.13.7 decan-1-amine sulfate, tricyclo 3.3.1.13,7 decan-1-amine, sulfate, tricyclo 3.3.1.13,7 decan-1-amine, sulfate 1:?, amantadine; sulfuric acid, 1-adamantanaminesulfate

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 200.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.