Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine 98.0+%, TCI America™
SDP

Catalog No. A231825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A231825G 25 g
A23185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A231825G Supplier TCI America Supplier No. A231825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N

Specifications

Melting Point 209°C
Color White
Quantity 25 g
UN Number 3259
Formula Weight 151.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanamine
RTECSNumber YD1925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.