Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Adamantaneacetic acid | 4942-47-6 | 194.27 | 25 G
SDP

Catalog No. 5000449781
Encompass_Preferred

1-Adamantaneacetic acid is an aliphatic carboxylic acid that can be used as an acylating agent, commonly found in chemical synthesis.

  • White to off-white solid
  • Stable under recommended storage conditions
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Melting point: 134-137°C
  • Boiling point: 319.2°C
  • Flash point: 163°C

Catalog No. 50-004-49781 Supplier Medchemexpress LLC Supplier No. 25GHYW001578
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Adamantaneacetic acid is an aliphatic carboxylic acid that can be used as an acylating agent, commonly found in chemical synthesis.

  • White to off-white solid
  • Stable under recommended storage conditions
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Melting point: 134-137°C
  • Boiling point: 319.2°C
  • Flash point: 163°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.