Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantaneacetonitrile, 97%

Catalog No. p-7027765
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2477003 1 g
AAB2477006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2477003 Supplier Thermo Scientific Chemicals Supplier No. B2477003
Only null left
Add to Cart

CAS: 16269-13-9 | C12H17N | 175.28 g/mol

1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16269-13-9
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00194034
InChI Key DXQVPXCZIRQITG-UHFFFAOYSA-N
Synonym 2-adamantan-1-yl acetonitrile, 1-adamantaneacetonitrile, 1-adamantylacetonitrile, 2-1-adamantyl acetonitrile, tricyclo 3.3.1.13,7 decane-1-acetonitrile, 2-adamantanylethanenitrile, pubchem21521, 1-adamantylmethyl cyanide, adamantane-1-acetonitrile
PubChem CID 140052
SMILES N#CCC12CC3CC(CC(C3)C1)C2

Specifications

Quantity 1 g
UN Number UN3439
Solubility Information Sparingly soluble in water.
IUPAC Name 2-(1-adamantyl)acetonitrile
Formula Weight 175.28
Percent Purity 97%
Chemical Name or Material 1-Adamantaneacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.