Learn More
Medchemexpress LLC 1-adamantyl (5-bromo-2-methoxybenzyl) amine | 332108-65-3 | 98.5% | 350.29 | C18H24BrNO | 100MG
SDP
Catalog No. 5000235107
Encompass_Preferred
ABMA is a small-molecule broad-spectrum inhibitor of intracellular toxins and pathogens that protects host cells by selectively targeting late endosome function. It is used in research to study host-pathogen interactions, endosomal trafficking, and intracellular infection mechanisms. Available as a high-purity solid and as DMSO solutions for cell-based assays.
  • High purity 98.5%.
  • Molecular formula C18H24BrNO; molecular weight 350.29.
  • Active against viruses, intracellular bacteria, and parasites.
  • Acts on host cell late endosomes rather than the pathogen.
  • Available as solid packs and 10 mM DMSO solutions for ready use.
Catalog No. 50-002-35107 Supplier Medchemexpress LLC Supplier No. HY124801100MG
Add to Cart
Add to Cart
ABMA is a small-molecule broad-spectrum inhibitor of intracellular toxins and pathogens that protects host cells by selectively targeting late endosome function. It is used in research to study host-pathogen interactions, endosomal trafficking, and intracellular infection mechanisms. Available as a high-purity solid and as DMSO solutions for cell-based assays.
  • High purity 98.5%.
  • Molecular formula C18H24BrNO; molecular weight 350.29.
  • Active against viruses, intracellular bacteria, and parasites.
  • Acts on host cell late endosomes rather than the pathogen.
  • Available as solid packs and 10 mM DMSO solutions for ready use.